The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032043.2 | NP_114432.2 | 19 - 20 | 3209 | 123 |
XM_011525332.2 | XP_011523634.1 | 20 - 21 | 3700 | 123 | |
XM_011525333.2 | XP_011523635.1 | 20 - 21 | 3697 | 123 | |
XM_011525334.2 | XP_011523636.1 | 20 - 21 | 4357 | 123 | |
XM_011525335.2 | XP_011523637.1 | 20 - 21 | 3640 | 123 | |
XM_011525336.2 | XP_011523638.1 | 19 - 20 | 3578 | 123 | |
XM_011525337.2 | XP_011523639.1 | 18 - 19 | 3497 | 123 | |
XM_011525338.2 | XP_011523640.1 | 16 - 17 | 2577 | 123 | |
XM_017025200.1 | XP_016880689.1 | 15 - 16 | 2517 | 123 | |
XM_017025201.1 | XP_016880690.1 | 15 - 16 | 3282 | 123 | |
XM_017025202.1 | XP_016880691.1 | 7 - 8 | 1645 | 123 | |
XM_017025203.1 | XP_016880692.1 | 8 - 9 | 1157 | 123 | |
GenBank mRNA | AF360549.1 | - | 19 - 20 | 3044 | 123 |
BC101472.1 | - | 19 - 20 | 2959 | 123 | |
BC101474.1 | - | 19 - 20 | 2959 | 123 |