The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017781.2 | NP_060251.2 | 3 - 4 | 502 | 80 |
XM_011515440.2 | XP_011513742.1 | 3 - 4 | 543 | 80 | |
XM_011515441.2 | XP_011513743.1 | 3 - 4 | 544 | 80 | |
GenBank mRNA | AK000366.1 | - | 3 - 4 | 343 | 80 |
BC025761.1 | - | 3 - 4 | 502 | 80 |