The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005026.3 | NP_005017.3 | 20 - 21 | 2804 | 61 |
XM_006710687.2 | XP_006710750.1 | 20 - 21 | 2864 | 61 | |
XM_006710689.2 | XP_006710752.1 | 19 - 20 | 2718 | 61 | |
XM_017001476.1 | XP_016856965.1 | 20 - 21 | 3087 | 61 | |
XM_017001477.1 | XP_016856966.1 | 21 - 22 | 3024 | 61 | |
XM_017001478.1 | XP_016856967.1 | 21 - 22 | 3184 | 61 | |
XM_017001479.1 | XP_016856968.1 | 20 - 21 | 3053 | 61 | |
XM_017001480.1 | XP_016856969.1 | 19 - 20 | 2895 | 61 | |
XM_017001481.1 | XP_016856970.1 | 19 - 20 | 2968 | 61 | |
XM_017001482.1 | XP_016856971.1 | 19 - 20 | 2890 | 61 | |
XM_017001483.1 | XP_016856972.1 | 19 - 20 | 2793 | 61 | |
XM_017001484.1 | XP_016856973.1 | 20 - 21 | 2907 | 61 | |
XM_017001485.1 | XP_016856974.1 | 19 - 20 | 2616 | 61 | |
GenBank mRNA | AB208835.1 | - | 15 - 16 | 1999 | 61 |
AK314191.1 | - | 20 - 21 | 2805 | 61 | |
BC132919.1 | - | 19 - 20 | 2718 | 61 | |
BC132921.1 | - | 19 - 20 | 2718 | 61 | |
BC144264.1 | - | 19 - 20 | 2715 | 61 | |
BC150297.1 | - | 20 - 21 | 2751 | 61 | |
DQ263594.1 | - | 18 - 19 | 2596 | 61 | |
DQ263595.1 | - | 18 - 19 | 2596 | 61 | |
DQ263596.1 | - | 18 - 19 | 2596 | 61 | |
JN190435.1 | - | 19 - 20 | 2831 | 61 | |
U57843.1 | - | 19 - 20 | 2783 | 61 | |
U86453.1 | - | 20 - 21 | 2791 | 61 | |
Y10055.2 | - | 20 - 21 | 2792 | 61 |