The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031748.3 | NP_001026918.2 | 2 - 3 | 226 | 99 |
NM_001319247.1 | NP_001306176.1 | 2 - 3 | 226 | 99 | |
GenBank mRNA | AL705293.1 | - | 2 - 3 | 202 | 99 |
BC038754.1 | - | 2 - 3 | 223 | 99 |