Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
Chromosome Location
Chr.22: 24719034 - 24774720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008496.3 NP_001008496.2 7 - 8 1196 145
NM_001255975.1 NP_001242904.1 7 - 8 1196 145
NR_045648.1 8 - 9 1406 145
NR_045649.1 8 - 9 1279 145
GenBank mRNA AB079368.1 - 7 - 8 1192 145
AK302409.1 - 7 - 8 1196 145
AK315806.1 - 7 - 8 1196 145
BC144693.1 - 8 - 9 1036 145
BC144696.1 - 8 - 9 1163 145

View Full Product Details