Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLB
Entrez Gene ID
868
Gene Name
Cbl proto-oncogene B
Gene Aliases
Cbl-b, Nbla00127, RNF56
Chromosome Location
Chr.3: 105655461 - 105869552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321786.1 NP_001308715.1 10 - 11 1640 63
NM_001321788.1 NP_001308717.1 10 - 11 1559 63
NM_001321789.1 NP_001308718.1 10 - 11 1578 63
NM_001321790.1 NP_001308719.1 10 - 11 1513 63
NM_001321791.1 NP_001308720.1 10 - 11 1559 63
NM_001321793.1 NP_001308722.1 10 - 11 1735 63
NM_001321794.1 NP_001308723.1 10 - 11 1559 63
NM_001321795.1 NP_001308724.1 10 - 11 1735 63
NM_001321796.1 NP_001308725.1 9 - 10 1412 63
NM_001321797.1 NP_001308726.1 10 - 11 1559 63
NM_001321798.1 NP_001308727.1 10 - 11 1735 63
NM_001321799.1 NP_001308728.1 9 - 10 1588 63
NM_001321806.1 NP_001308735.1 9 - 10 1402 63
NM_001321807.1 NP_001308736.1 11 - 12 1879 63
NM_001321808.1 NP_001308737.1 11 - 12 1703 63
NM_001321811.1 NP_001308740.1 9 - 10 1578 63
NM_001321813.1 NP_001308742.1 11 - 12 1616 63
NM_001321816.1 NP_001308745.1 9 - 10 1578 63
NM_001321820.1 NP_001308749.1 12 - 13 1867 63
NM_001321822.1 NP_001308751.1 11 - 12 1785 63
NM_170662.4 NP_733762.2 10 - 11 1735 63
NR_135806.1 11 - 12 1867 63
NR_135807.1 11 - 12 1783 63
NR_135808.1 10 - 11 1735 63
NR_135810.1 10 - 11 1513 63
NR_135811.1 10 - 11 1559 63
NR_135812.1 10 - 11 1455 63
XM_011513257.1 XP_011511559.1 10 - 11 1555 63
XM_011513259.2 XP_011511561.2 10 - 11 1680 63
XM_017007395.1 XP_016862884.1 10 - 11 1507 63
XM_017007396.1 XP_016862885.1 10 - 11 1678 63
XM_017007397.1 XP_016862886.1 9 - 10 1532 63
XM_017007398.1 XP_016862887.1 9 - 10 1358 63
XM_017007399.1 XP_016862888.1 9 - 10 1583 63
XM_017007400.1 XP_016862889.1 9 - 10 1532 63
GenBank mRNA AK292792.1 - 10 - 11 1676 63
AK302533.1 - 10 - 11 1513 63
AK316276.1 - 10 - 11 1493 63
BC032851.2 - 10 - 11 1676 63
DQ349203.1 - 9 - 10 1413 63
DQ349204.1 - 9 - 10 1413 63
U26710.1 - 10 - 11 1735 63
U26711.1 - 10 - 11 1735 63
U26712.1 - 10 - 11 1735 63

View Full Product Details