The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321793.1 | NP_001308722.1 | 1 - 2 | 311 | 118 |
NM_001321795.1 | NP_001308724.1 | 1 - 2 | 311 | 118 | |
NM_001321798.1 | NP_001308727.1 | 1 - 2 | 311 | 118 | |
NM_001321799.1 | NP_001308728.1 | 1 - 2 | 311 | 118 | |
NM_001321807.1 | NP_001308736.1 | 1 - 2 | 311 | 118 | |
NM_001321811.1 | NP_001308740.1 | 1 - 2 | 311 | 118 | |
NM_001321816.1 | NP_001308745.1 | 1 - 2 | 311 | 118 | |
NM_001321822.1 | NP_001308751.1 | 1 - 2 | 311 | 118 | |
NM_170662.4 | NP_733762.2 | 1 - 2 | 311 | 118 | |
NR_135806.1 | 1 - 2 | 311 | 118 | ||
NR_135807.1 | 1 - 2 | 311 | 118 | ||
NR_135808.1 | 1 - 2 | 311 | 118 | ||
NR_135809.1 | 1 - 2 | 311 | 118 | ||
XM_017007399.1 | XP_016862888.1 | 1 - 2 | 306 | 118 | |
GenBank mRNA | AK094184.1 | - | 1 - 2 | 252 | 118 |
AK292792.1 | - | 1 - 2 | 252 | 118 | |
BC032851.2 | - | 1 - 2 | 252 | 118 | |
U26710.1 | - | 1 - 2 | 311 | 118 | |
U26711.1 | - | 1 - 2 | 311 | 118 | |
U26712.1 | - | 1 - 2 | 311 | 118 |