Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CBLB
- Entrez Gene ID
-
868
- Gene Name
- Cbl proto-oncogene B
- Gene Aliases
-
Cbl-b, Nbla00127, RNF56
- Chromosome Location
-
Chr.3: 105655461 - 105869552 on Build GRCh38
View Full Product Details