Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLB
Entrez Gene ID
868
Gene Name
Cbl proto-oncogene B
Gene Aliases
Cbl-b, Nbla00127, RNF56
Chromosome Location
Chr.3: 105655461 - 105869552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321786.1 NP_001308715.1 7 - 8 1213 113
NM_001321788.1 NP_001308717.1 7 - 8 1132 113
NM_001321789.1 NP_001308718.1 7 - 8 1151 113
NM_001321790.1 NP_001308719.1 7 - 8 1086 113
NM_001321791.1 NP_001308720.1 7 - 8 1132 113
NM_001321793.1 NP_001308722.1 7 - 8 1308 113
NM_001321794.1 NP_001308723.1 7 - 8 1132 113
NM_001321795.1 NP_001308724.1 7 - 8 1308 113
NM_001321796.1 NP_001308725.1 6 - 7 985 113
NM_001321797.1 NP_001308726.1 7 - 8 1132 113
NM_001321798.1 NP_001308727.1 7 - 8 1308 113
NM_001321799.1 NP_001308728.1 6 - 7 1161 113
NM_001321806.1 NP_001308735.1 6 - 7 975 113
NM_001321807.1 NP_001308736.1 8 - 9 1452 113
NM_001321808.1 NP_001308737.1 8 - 9 1276 113
NM_001321811.1 NP_001308740.1 6 - 7 1151 113
NM_001321813.1 NP_001308742.1 8 - 9 1189 113
NM_001321816.1 NP_001308745.1 6 - 7 1151 113
NM_001321820.1 NP_001308749.1 8 - 9 1276 113
NM_001321822.1 NP_001308751.1 7 - 8 1308 113
NM_170662.4 NP_733762.2 7 - 8 1308 113
NR_135806.1 8 - 9 1440 113
NR_135807.1 8 - 9 1356 113
NR_135808.1 7 - 8 1308 113
NR_135809.1 7 - 8 1308 113
NR_135810.1 7 - 8 1086 113
NR_135811.1 7 - 8 1132 113
NR_135812.1 7 - 8 1028 113
XM_011513257.1 XP_011511559.1 7 - 8 1128 113
XM_011513259.2 XP_011511561.2 7 - 8 1253 113
XM_017007395.1 XP_016862884.1 7 - 8 1080 113
XM_017007396.1 XP_016862885.1 7 - 8 1251 113
XM_017007397.1 XP_016862886.1 6 - 7 1105 113
XM_017007398.1 XP_016862887.1 6 - 7 931 113
XM_017007399.1 XP_016862888.1 6 - 7 1156 113
XM_017007400.1 XP_016862889.1 6 - 7 1105 113
GenBank mRNA AK292792.1 - 7 - 8 1249 113
AK302533.1 - 7 - 8 1086 113
AK302892.1 - 7 - 8 1061 113
AK316276.1 - 7 - 8 1066 113
BC032851.2 - 7 - 8 1249 113
DQ349203.1 - 6 - 7 986 113
DQ349204.1 - 6 - 7 986 113
U26710.1 - 7 - 8 1308 113
U26711.1 - 7 - 8 1308 113
U26712.1 - 7 - 8 1308 113

View Full Product Details