The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003826.2 | NP_003817.1 | 1 - 2 | 284 | 72 |
GenBank mRNA | AK314495.1 | - | 1 - 2 | 123 | 72 |
BC001889.1 | - | 1 - 2 | 120 | 72 | |
CR536554.1 | - | 1 - 2 | 57 | 72 | |
DA296597.1 | - | 1 - 2 | 303 | 72 | |
U78107.1 | - | 1 - 2 | 130 | 72 |