The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040613.2 | NP_001035703.1 | 1 - 2 | 349 | 131 |
NM_017866.5 | NP_060336.3 | 1 - 2 | 349 | 131 | |
NR_033334.1 | 1 - 2 | 349 | 131 | ||
GenBank mRNA | AI800622.1 | - | 1 - 2 | 137 | 131 |
BC002748.2 | - | 1 - 2 | 283 | 131 | |
BF439499.1 | - | 1 - 2 | 119 | 131 | |
BU542365.1 | - | 1 - 2 | 271 | 131 | |
CB961613.1 | - | 1 - 2 | 323 | 131 | |
DB483583.1 | - | 1 - 2 | 351 | 131 |