The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257376.1 | NP_001244305.1 | 10 - 11 | 1012 | 104 |
NM_001283031.1 | NP_001269960.1 | 11 - 12 | 1086 | 104 | |
NM_001283032.1 | NP_001269961.1 | 11 - 12 | 1085 | 104 | |
NM_001283033.1 | NP_001269962.1 | 9 - 10 | 872 | 104 | |
NM_016106.3 | NP_057190.2 | 12 - 13 | 1174 | 104 | |
NM_182835.2 | NP_878255.1 | 11 - 12 | 1103 | 104 | |
XM_005267469.2 | XP_005267526.1 | 12 - 13 | 1172 | 104 | |
XM_011536586.2 | XP_011534888.1 | 12 - 13 | 1171 | 104 | |
XM_011536587.2 | XP_011534889.1 | 12 - 13 | 1171 | 104 | |
XM_011536588.2 | XP_011534890.1 | 12 - 13 | 1171 | 104 | |
XM_011536589.1 | XP_011534891.1 | 12 - 13 | 1171 | 104 | |
XM_017021113.1 | XP_016876602.1 | 12 - 13 | 1171 | 104 | |
GenBank mRNA | AB020724.2 | - | 12 - 13 | 1109 | 104 |
AF086916.1 | - | 12 - 13 | 1097 | 104 | |
AF092139.1 | - | 12 - 13 | 1090 | 104 | |
AF110646.1 | - | 12 - 13 | 1085 | 104 | |
AF319958.1 | - | 12 - 13 | 1113 | 104 | |
AK222820.1 | - | 12 - 13 | 1093 | 104 | |
AK290410.1 | - | 12 - 13 | 1109 | 104 | |
AK297873.1 | - | 11 - 12 | 1038 | 104 | |
AK298622.1 | - | 10 - 11 | 999 | 104 | |
AK299222.1 | - | 11 - 12 | 1013 | 104 | |
AK301406.1 | - | 11 - 12 | 1020 | 104 | |
AK307960.1 | - | 9 - 10 | 806 | 104 | |
AK308587.1 | - | 11 - 12 | 1042 | 104 | |
AK316212.1 | - | 11 - 12 | 1038 | 104 | |
BC017734.1 | - | 12 - 13 | 1089 | 104 |