The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257376.1 | NP_001244305.1 | 13 - 14 | 1268 | 119 |
NM_001283031.1 | NP_001269960.1 | 14 - 15 | 1342 | 119 | |
NM_001283032.1 | NP_001269961.1 | 14 - 15 | 1341 | 119 | |
NM_001283033.1 | NP_001269962.1 | 12 - 13 | 1128 | 119 | |
NM_016106.3 | NP_057190.2 | 15 - 16 | 1430 | 119 | |
NM_182835.2 | NP_878255.1 | 14 - 15 | 1359 | 119 | |
XM_005267469.2 | XP_005267526.1 | 15 - 16 | 1428 | 119 | |
XM_011536586.2 | XP_011534888.1 | 15 - 16 | 1427 | 119 | |
XM_011536587.2 | XP_011534889.1 | 15 - 16 | 1427 | 119 | |
GenBank mRNA | AB020724.2 | - | 15 - 16 | 1365 | 119 |
AF086916.1 | - | 15 - 16 | 1353 | 119 | |
AF092139.1 | - | 15 - 16 | 1346 | 119 | |
AF110646.1 | - | 15 - 16 | 1341 | 119 | |
AF319958.1 | - | 15 - 16 | 1369 | 119 | |
AK222820.1 | - | 15 - 16 | 1349 | 119 | |
AK290410.1 | - | 15 - 16 | 1365 | 119 | |
AK297873.1 | - | 14 - 15 | 1294 | 119 | |
AK298622.1 | - | 13 - 14 | 1255 | 119 | |
AK299222.1 | - | 14 - 15 | 1269 | 119 | |
AK301406.1 | - | 14 - 15 | 1276 | 119 | |
AK307960.1 | - | 12 - 13 | 1062 | 119 | |
AK308587.1 | - | 14 - 15 | 1298 | 119 | |
AK316212.1 | - | 14 - 15 | 1294 | 119 | |
BC017734.1 | - | 15 - 16 | 1345 | 119 |