The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257376.1 | NP_001244305.1 | 2 - 3 | 232 | 152 |
NM_001283031.1 | NP_001269960.1 | 4 - 5 | 394 | 152 | |
NM_001283032.1 | NP_001269961.1 | 3 - 4 | 305 | 152 | |
NM_016106.3 | NP_057190.2 | 4 - 5 | 394 | 152 | |
NM_182835.2 | NP_878255.1 | 3 - 4 | 323 | 152 | |
XM_005267469.2 | XP_005267526.1 | 4 - 5 | 392 | 152 | |
XM_011536586.2 | XP_011534888.1 | 4 - 5 | 391 | 152 | |
XM_011536587.2 | XP_011534889.1 | 4 - 5 | 391 | 152 | |
XM_011536588.2 | XP_011534890.1 | 4 - 5 | 391 | 152 | |
XM_011536589.1 | XP_011534891.1 | 4 - 5 | 391 | 152 | |
XM_017021113.1 | XP_016876602.1 | 4 - 5 | 391 | 152 | |
GenBank mRNA | AB020724.2 | - | 4 - 5 | 329 | 152 |
AF086916.1 | - | 4 - 5 | 317 | 152 | |
AF092139.1 | - | 4 - 5 | 310 | 152 | |
AF110646.1 | - | 4 - 5 | 305 | 152 | |
AF319958.1 | - | 4 - 5 | 333 | 152 | |
AK222820.1 | - | 4 - 5 | 313 | 152 | |
AK290410.1 | - | 4 - 5 | 329 | 152 | |
AK297873.1 | - | 3 - 4 | 258 | 152 | |
AK298622.1 | - | 2 - 3 | 219 | 152 | |
AK299222.1 | - | 4 - 5 | 321 | 152 | |
AK301406.1 | - | 3 - 4 | 240 | 152 | |
AK316212.1 | - | 3 - 4 | 258 | 152 | |
BC017734.1 | - | 4 - 5 | 309 | 152 | |
BG028643.1 | - | 3 - 4 | 236 | 152 | |
BU198450.1 | - | 4 - 5 | 152 | ||
CA454055.1 | - | 4 - 5 | 320 | 152 | |
DB105324.1 | - | 3 - 4 | 323 | 152 |