The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003551.2 | NP_003542.1 | 1 - 2 | 77 | 75 |
GenBank mRNA | AF067724.1 | - | 1 - 2 | 47 | 75 |
BC026182.2 | - | 1 - 2 | 49 | 75 |