The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016429.3 | NP_057513.1 | 2 - 3 | 215 | 101 |
XM_011524874.2 | XP_011523176.1 | 2 - 3 | 214 | 101 | |
GenBank mRNA | AB037938.1 | - | 2 - 3 | 199 | 101 |
AB047849.1 | - | 2 - 3 | 197 | 101 | |
BC015924.1 | - | 2 - 3 | 191 | 101 | |
HY179834.1 | - | 2 - 3 | 210 | 101 |