Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPZ2
Entrez Gene ID
51226
Gene Name
coatomer protein complex subunit zeta 2
Gene Aliases
zeta2-COP
Chromosome Location
Chr.17: 48026167 - 48048091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016429.3 NP_057513.1 5 - 6 440 62
GenBank mRNA AB037938.1 - 5 - 6 424 62
AB047849.1 - 5 - 6 422 62
BC015924.1 - 5 - 6 416 62

View Full Product Details