The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037333.2 | NP_001032410.1 | 23 - 24 | 2817 | 67 |
NM_001291721.1 | NP_001278650.1 | 22 - 23 | 2739 | 67 | |
NM_001291722.1 | NP_001278651.1 | 24 - 25 | 2892 | 67 | |
NM_014376.3 | NP_055191.2 | 23 - 24 | 2943 | 67 | |
XM_011534516.2 | XP_011532818.1 | 23 - 24 | 3083 | 67 | |
XM_017009341.1 | XP_016864830.1 | 23 - 24 | 2932 | 67 | |
XM_017009342.1 | XP_016864831.1 | 24 - 25 | 2854 | 67 | |
GenBank mRNA | AB032994.2 | - | 24 - 25 | 2832 | 67 |
AF160973.1 | - | 23 - 24 | 2726 | 67 | |
AK223604.1 | - | 23 - 24 | 2770 | 67 | |
AK294220.1 | - | 18 - 19 | 2186 | 67 | |
AK295865.1 | - | 20 - 21 | 2366 | 67 | |
AK297401.1 | - | 21 - 22 | 2549 | 67 | |
AK303702.1 | - | 22 - 23 | 2692 | 67 | |
AL136549.1 | - | 23 - 24 | 2816 | 67 | |
AL161999.1 | - | 15 - 16 | 1777 | 67 | |
BC011762.1 | - | 23 - 24 | 2943 | 67 | |
BC021008.1 | - | 23 - 24 | 2943 | 67 | |
L47738.1 | - | 12 - 13 | 1592 | 67 |