The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144650.2 | NP_653251.2 | 5 - 6 | 385 | 80 |
GenBank mRNA | AK056992.1 | - | 4 - 5 | 322 | 80 |
AK293530.1 | - | 2 - 3 | 225 | 80 | |
AK300243.1 | - | 5 - 6 | 364 | 80 | |
AY033237.1 | - | 5 - 6 | 370 | 80 |