The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288705.1 | NP_001275634.1 | 13 - 14 | 2187 | 65 |
NM_005211.3 | NP_005202.2 | 14 - 15 | 2266 | 65 | |
GenBank mRNA | BC047521.1 | - | 13 - 14 | 2169 | 65 |
BX648599.1 | - | 12 - 13 | 2057 | 65 | |
X03663.1 | - | 14 - 15 | 2274 | 65 |