The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135110.2 | NP_001128582.1 | 5 - 6 | 860 | 74 |
NM_001286516.1 | NP_001273445.1 | 3 - 4 | 511 | 74 | |
NM_005147.5 | NP_005138.3 | 5 - 6 | 860 | 74 | |
GenBank mRNA | AB209923.1 | - | 5 - 6 | 779 | 74 |
AF061749.1 | - | 5 - 6 | 814 | 74 | |
AF244136.1 | - | 5 - 6 | 785 | 74 | |
AF411044.1 | - | 5 - 6 | 827 | 74 | |
AK001333.1 | - | 5 - 6 | 800 | 74 | |
AK223105.1 | - | 5 - 6 | 800 | 74 | |
AK295391.1 | - | 3 - 4 | 451 | 74 | |
AK314218.1 | - | 5 - 6 | 797 | 74 | |
BC007225.1 | - | 5 - 6 | 780 | 74 | |
BC011855.2 | - | 5 - 6 | 784 | 74 | |
BC012343.1 | - | 5 - 6 | 776 | 74 | |
BC014062.2 | - | 5 - 6 | 776 | 74 | |
BC020248.1 | - | 1 - 2 | 109 | 74 | |
BC030145.2 | - | 5 - 6 | 776 | 74 | |
BC032100.2 | - | 5 - 6 | 798 | 74 |