The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007284.3 | NP_009215.1 | 2 - 3 | 251 | 80 |
GenBank mRNA | AF109365.1 | - | 2 - 3 | 152 | 80 |
AL136773.1 | - | 2 - 3 | 218 | 80 | |
BC000327.2 | - | 2 - 3 | 202 | 80 | |
BC003161.1 | - | 2 - 3 | 168 | 80 | |
BC016452.1 | - | 2 - 3 | 218 | 80 | |
CR456732.1 | - | 2 - 3 | 100 | 80 | |
CR533520.1 | - | 2 - 3 | 100 | 80 | |
Y17169.1 | - | 2 - 3 | 204 | 80 |