The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010972.1 | NP_001010972.1 | 7 - 8 | 1469 | 84 |
NM_003461.4 | NP_003452.1 | 7 - 8 | 1472 | 84 | |
XM_011516569.2 | XP_011514871.2 | 6 - 7 | 1367 | 84 | |
XM_017012587.1 | XP_016868076.1 | 7 - 8 | 1562 | 84 | |
GenBank mRNA | AK130138.1 | - | 3 - 4 | 762 | 84 |
AK298925.1 | - | 5 - 6 | 1218 | 84 | |
AK299005.1 | - | 6 - 7 | 1188 | 84 | |
AK302309.1 | - | 6 - 7 | 1280 | 84 | |
AK316227.1 | - | 6 - 7 | 1164 | 84 | |
BC002323.2 | - | 7 - 8 | 1341 | 84 | |
BC008743.2 | - | 7 - 8 | 1436 | 84 | |
BC009360.2 | - | 7 - 8 | 1381 | 84 | |
BC010031.2 | - | 7 - 8 | 1369 | 84 | |
CR457431.1 | - | 6 - 7 | 1314 | 84 | |
U15158.1 | - | 5 - 6 | 1077 | 84 | |
X94991.1 | - | 6 - 7 | 1324 | 84 | |
X95735.1 | - | 7 - 8 | 1385 | 84 |