The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001248003.1 | NP_001234932.1 | 1 - 2 | 281 | 69 |
NM_001248005.1 | NP_001234934.1 | 1 - 2 | 281 | 69 | |
XM_006719112.3 | XP_006719175.1 | 1 - 2 | 249 | 69 | |
XM_011520766.2 | XP_011519068.1 | 1 - 2 | 249 | 69 | |
XM_011520768.2 | XP_011519070.1 | 1 - 2 | 250 | 69 | |
XM_011520769.2 | XP_011519071.1 | 1 - 2 | 249 | 69 | |
XM_017019666.1 | XP_016875155.1 | 1 - 2 | 215 | 69 | |
GenBank mRNA | AF246962.1 | - | 1 - 2 | 80 | 69 |
AK296706.1 | - | 1 - 2 | 215 | 69 | |
BC125062.1 | - | 1 - 2 | 80 | 69 |