The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207062.1 | NP_001193991.1 | 4 - 5 | 484 | 77 |
NM_032561.4 | NP_115950.3 | 5 - 6 | 695 | 77 | |
XM_005261781.1 | XP_005261838.1 | 5 - 6 | 659 | 77 | |
XM_005261782.3 | XP_005261839.1 | 5 - 6 | 539 | 77 | |
XM_005261783.2 | XP_005261840.1 | 4 - 5 | 596 | 77 | |
XM_005261784.2 | XP_005261841.1 | 4 - 5 | 596 | 77 | |
GenBank mRNA | AF324466.1 | - | 4 - 5 | 495 | 77 |
AK057349.1 | - | 5 - 6 | 745 | 77 | |
BC031998.1 | - | 5 - 6 | 577 | 77 | |
BI827388.1 | - | 4 - 5 | 484 | 77 | |
CR456440.1 | - | 5 - 6 | 575 | 77 |