The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319035.1 | NP_001305964.1 | 48 - 49 | 6935 | 144 |
NM_007110.4 | NP_009041.2 | 50 - 51 | 7259 | 144 | |
XM_005268027.4 | XP_005268084.1 | 50 - 51 | 7399 | 144 | |
XM_011537110.2 | XP_011535412.1 | 42 - 43 | 5744 | 144 | |
GenBank mRNA | AB209669.1 | - | 38 - 39 | 5245 | 144 |
BC126107.1 | - | 50 - 51 | 7259 | 144 | |
BC143812.1 | - | 48 - 49 | 7124 | 144 | |
BX640983.1 | - | 3 - 4 | 358 | 144 | |
U86136.2 | - | 50 - 51 | 7259 | 144 |