The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 24 - 25 | 2598 | 65 |
GenBank mRNA | AB208897.1 | - | 24 - 25 | 2487 | 65 |
AF411122.1 | - | 24 - 25 | 2469 | 65 | |
AK026991.1 | - | 17 - 18 | 1773 | 65 | |
AK027871.1 | - | 6 - 7 | 566 | 65 | |
AK056702.1 | - | 24 - 25 | 2445 | 65 | |
AK057044.1 | - | 23 - 24 | 2579 | 65 | |
AK057277.1 | - | 24 - 25 | 2502 | 65 | |
AK074743.1 | - | 9 - 10 | 857 | 65 | |
AK094107.1 | - | 24 - 25 | 2446 | 65 | |
AK094897.1 | - | 23 - 24 | 2605 | 65 | |
AK314305.1 | - | 22 - 23 | 2470 | 65 | |
BC003690.1 | - | 15 - 16 | 1585 | 65 | |
BC110804.1 | - | 24 - 25 | 2500 | 65 | |
BC136759.1 | - | 24 - 25 | 2549 | 65 | |
BX248267.1 | - | 24 - 25 | 2480 | 65 | |
CR457367.1 | - | 15 - 16 | 1495 | 65 |