The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 25 - 26 | 2729 | 66 |
NR_051979.1 | 25 - 26 | 2749 | 66 | ||
GenBank mRNA | AB208897.1 | - | 25 - 26 | 2618 | 66 |
AF411122.1 | - | 25 - 26 | 2600 | 66 | |
AK027871.1 | - | 7 - 8 | 697 | 66 | |
AK056527.1 | - | 25 - 26 | 2670 | 66 | |
AK056702.1 | - | 25 - 26 | 2576 | 66 | |
AK057044.1 | - | 24 - 25 | 2710 | 66 | |
AK057277.1 | - | 25 - 26 | 2633 | 66 | |
AK074743.1 | - | 10 - 11 | 988 | 66 | |
AK094107.1 | - | 25 - 26 | 2577 | 66 | |
AK094897.1 | - | 24 - 25 | 2736 | 66 | |
AK095306.1 | - | 25 - 26 | 2577 | 66 | |
AK302097.1 | - | 19 - 20 | 1787 | 66 | |
AK314305.1 | - | 23 - 24 | 2601 | 66 | |
BC003690.1 | - | 16 - 17 | 1716 | 66 | |
BC110804.1 | - | 25 - 26 | 2631 | 66 | |
BC136759.1 | - | 25 - 26 | 2680 | 66 | |
BC144399.1 | - | 25 - 26 | 2700 | 66 | |
BX248267.1 | - | 25 - 26 | 2611 | 66 | |
CR457367.1 | - | 16 - 17 | 1626 | 66 |