The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 26 - 27 | 2939 | 91 |
NR_051979.1 | 26 - 27 | 2959 | 91 | ||
GenBank mRNA | AB208897.1 | - | 26 - 27 | 2828 | 91 |
AF411122.1 | - | 26 - 27 | 2810 | 91 | |
AK026991.1 | - | 19 - 20 | 2114 | 91 | |
AK056527.1 | - | 26 - 27 | 2880 | 91 | |
AK056702.1 | - | 26 - 27 | 2786 | 91 | |
AK057044.1 | - | 25 - 26 | 2920 | 91 | |
AK057277.1 | - | 26 - 27 | 2843 | 91 | |
AK074743.1 | - | 11 - 12 | 1198 | 91 | |
AK094107.1 | - | 26 - 27 | 2787 | 91 | |
AK094897.1 | - | 25 - 26 | 2946 | 91 | |
AK095306.1 | - | 26 - 27 | 2787 | 91 | |
AK302097.1 | - | 20 - 21 | 1997 | 91 | |
AK314305.1 | - | 24 - 25 | 2811 | 91 | |
BC003690.1 | - | 17 - 18 | 1926 | 91 | |
BC110804.1 | - | 26 - 27 | 2841 | 91 | |
BC136759.1 | - | 26 - 27 | 2890 | 91 | |
BC144399.1 | - | 26 - 27 | 2910 | 91 | |
BX248267.1 | - | 26 - 27 | 2821 | 91 | |
CR457367.1 | - | 17 - 18 | 1836 | 91 |