The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 6 - 7 | 723 | 70 |
NR_051979.1 | 6 - 7 | 723 | 70 | ||
GenBank mRNA | AB208897.1 | - | 6 - 7 | 613 | 70 |
AF411122.1 | - | 6 - 7 | 594 | 70 | |
AK056527.1 | - | 6 - 7 | 644 | 70 | |
AK056702.1 | - | 6 - 7 | 570 | 70 | |
AK057044.1 | - | 5 - 6 | 681 | 70 | |
AK057277.1 | - | 6 - 7 | 627 | 70 | |
AK094107.1 | - | 6 - 7 | 572 | 70 | |
AK094897.1 | - | 5 - 6 | 730 | 70 | |
AK095306.1 | - | 6 - 7 | 571 | 70 | |
AK314305.1 | - | 6 - 7 | 602 | 70 | |
BC110804.1 | - | 6 - 7 | 625 | 70 | |
BC136759.1 | - | 6 - 7 | 674 | 70 | |
BC144399.1 | - | 6 - 7 | 674 | 70 | |
BX248267.1 | - | 6 - 7 | 605 | 70 |