Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
Chromosome Location
Chr.14: 24180219 - 24188918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024658.3 NP_078934.3 1 - 2 202 71
NR_051979.1 1 - 2 202 71
GenBank mRNA AB208897.1 - 1 - 2 92 71
AF411122.1 - 1 - 2 73 71
AK056527.1 - 1 - 2 106 71
AK056702.1 - 1 - 2 81 71
AK057277.1 - 1 - 2 106 71
AK094897.1 - 1 - 2 81 71
AK095306.1 - 1 - 2 105 71
AK302097.1 - 1 - 2 92 71
AK314305.1 - 1 - 2 81 71
BC110804.1 - 1 - 2 104 71
BC136759.1 - 1 - 2 153 71
BC144399.1 - 1 - 2 153 71
BG333193.1 - 1 - 2 202 71
BX248267.1 - 1 - 2 84 71

View Full Product Details