Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAZAP1
Entrez Gene ID
26528
Gene Name
DAZ associated protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 1407569 - 1435687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170711.2 NP_733829.1 11 - 12 1253 104
XM_011527908.2 XP_011526210.1 11 - 12 2525 104
XM_011527909.2 XP_011526211.1 11 - 12 2525 104
GenBank mRNA AK056850.1 - 9 - 10 876 104

View Full Product Details