Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAZAP1
Entrez Gene ID
26528
Gene Name
DAZ associated protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 1407569 - 1435687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005259531.3 XP_005259588.1 4 - 5 462 86
XM_005259535.3 XP_005259592.1 4 - 5 470 86
XM_011527906.2 XP_011526208.1 4 - 5 1782 86
XM_011527907.2 XP_011526209.1 4 - 5 1791 86
GenBank mRNA BU557183.1 - - 509 86

View Full Product Details