The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002703.4 | NP_002694.3 | 4 - 5 | 674 | 70 |
XM_011534387.1 | XP_011532689.1 | 4 - 5 | 451 | 70 | |
GenBank mRNA | AK222759.1 | - | 4 - 5 | 613 | 70 |
AK290942.1 | - | 4 - 5 | 529 | 70 | |
AK312297.1 | - | 4 - 5 | 650 | 70 | |
BC004200.2 | - | 4 - 5 | 546 | 70 | |
D13757.1 | - | 4 - 5 | 636 | 70 | |
U00238.1 | - | 4 - 5 | 565 | 70 |