The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002703.4 | NP_002694.3 | 9 - 10 | 1399 | 143 |
XM_011534387.1 | XP_011532689.1 | 9 - 10 | 1176 | 143 | |
GenBank mRNA | AB209246.1 | - | 1 - 2 | 1743 | 143 |
AK222759.1 | - | 9 - 10 | 1338 | 143 | |
AK290942.1 | - | 9 - 10 | 1254 | 143 | |
AK312297.1 | - | 9 - 10 | 1375 | 143 | |
BC004200.2 | - | 9 - 10 | 1271 | 143 | |
D13757.1 | - | 9 - 10 | 1361 | 143 | |
U00238.1 | - | 9 - 10 | 1290 | 143 |