The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308133.1 | NP_001295062.1 | 7 - 8 | 1412 | 145 |
NM_001308134.1 | NP_001295063.1 | 8 - 9 | 1490 | 145 | |
NM_020937.3 | NP_065988.1 | 8 - 9 | 1490 | 145 | |
XM_011537034.2 | XP_011535336.1 | 8 - 9 | 1489 | 145 | |
XM_011537035.2 | XP_011535337.1 | 7 - 8 | 1411 | 145 | |
XM_017021523.1 | XP_016877012.1 | 8 - 9 | 1489 | 145 | |
XM_017021524.1 | XP_016877013.1 | 6 - 7 | 711 | 145 | |
XM_017021525.1 | XP_016877014.1 | 6 - 7 | 1278 | 145 | |
XM_017021526.1 | XP_016877015.1 | 5 - 6 | 522 | 145 | |
XM_017021527.1 | XP_016877016.1 | 7 - 8 | 1303 | 145 | |
GenBank mRNA | AK093422.1 | - | 8 - 9 | 1489 | 145 |
AL833656.1 | - | 2 - 3 | 152 | 145 | |
BC036056.1 | - | 8 - 9 | 1491 | 145 | |
BC140776.1 | - | 7 - 8 | 1397 | 145 | |
BC144511.1 | - | 7 - 8 | 1397 | 145 | |
DQ140356.1 | - | 8 - 9 | 1497 | 145 |