Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FANCM
Entrez Gene ID
57697
Gene Name
Fanconi anemia complementation group M
Gene Aliases
FAAP250, KIAA1596
Chromosome Location
Chr.14: 45135939 - 45200890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308133.1 NP_001295062.1 15 - 16 4396 130
NM_020937.3 NP_065988.1 16 - 17 4474 130
XM_017021527.1 XP_016877016.1 15 - 16 4287 130
GenBank mRNA AK001672.1 - 3 - 4 1178 130
BC140776.1 - 15 - 16 4381 130
BC144511.1 - 15 - 16 4381 130
DQ140356.1 - 16 - 17 4481 130

View Full Product Details