The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173550.2 | NP_775821.2 | 22 - 23 | 3727 | 122 |
XM_005251397.2 | XP_005251454.1 | 22 - 23 | 3634 | 122 | |
XM_005251398.3 | XP_005251455.1 | 22 - 23 | 3873 | 122 | |
XM_005251399.4 | XP_005251456.1 | 23 - 24 | 3933 | 122 | |
XM_011517788.2 | XP_011516090.1 | 22 - 23 | 3863 | 122 | |
XM_011517791.2 | XP_011516093.1 | 21 - 22 | 3741 | 122 | |
XM_011517796.2 | XP_011516098.1 | 20 - 21 | 3280 | 122 | |
XM_017014431.1 | XP_016869920.1 | 22 - 23 | 3873 | 122 | |
XM_017014432.1 | XP_016869921.1 | 22 - 23 | 3873 | 122 | |
XM_017014433.1 | XP_016869922.1 | 22 - 23 | 3873 | 122 | |
XM_017014434.1 | XP_016869923.1 | 22 - 23 | 3873 | 122 | |
XM_017014435.1 | XP_016869924.1 | 22 - 23 | 3873 | 122 | |
XM_017014436.1 | XP_016869925.1 | 22 - 23 | 3873 | 122 | |
XM_017014437.1 | XP_016869926.1 | 22 - 23 | 3873 | 122 | |
XM_017014438.1 | XP_016869927.1 | 22 - 23 | 3872 | 122 | |
XM_017014439.1 | XP_016869928.1 | 23 - 24 | 4035 | 122 | |
XM_017014440.1 | XP_016869929.1 | 20 - 21 | 3294 | 122 | |
XM_017014441.1 | XP_016869930.1 | 22 - 23 | 3872 | 122 | |
XM_017014442.1 | XP_016869931.1 | 22 - 23 | 3872 | 122 | |
XM_017014443.1 | XP_016869932.1 | 21 - 22 | 3355 | 122 | |
XM_017014444.1 | XP_016869933.1 | 11 - 12 | 2074 | 122 | |
GenBank mRNA | AF440401.1 | - | 2 - 3 | 208 | 122 |
AK094506.1 | - | 4 - 5 | 573 | 122 | |
AK096586.1 | - | 6 - 7 | 1161 | 122 | |
AK128581.1 | - | 10 - 11 | 1990 | 122 | |
AY422473.1 | - | 22 - 23 | 3727 | 122 | |
BC026063.2 | - | 6 - 7 | 1002 | 122 | |
BC064849.1 | - | 9 - 10 | 1933 | 122 | |
BC144214.1 | - | 21 - 22 | 3528 | 122 | |
BC150600.1 | - | 21 - 22 | 3528 | 122 | |
CR936775.1 | - | 23 - 24 | 4005 | 122 |