Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC171
Entrez Gene ID
203238
Gene Name
coiled-coil domain containing 171
Gene Aliases
C9orf93, bA536D16.1, bA778P13.1
Chromosome Location
Chr.9: 15552874 - 16062431 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173550.2 NP_775821.2 14 - 15 2004 93
XM_005251397.2 XP_005251454.1 14 - 15 1911 93
XM_005251398.3 XP_005251455.1 14 - 15 2150 93
XM_005251399.4 XP_005251456.1 15 - 16 2210 93
XM_011517788.2 XP_011516090.1 14 - 15 2140 93
XM_011517791.2 XP_011516093.1 13 - 14 2018 93
XM_011517796.2 XP_011516098.1 12 - 13 1557 93
XM_017014431.1 XP_016869920.1 14 - 15 2150 93
XM_017014432.1 XP_016869921.1 14 - 15 2150 93
XM_017014433.1 XP_016869922.1 14 - 15 2150 93
XM_017014434.1 XP_016869923.1 14 - 15 2150 93
XM_017014435.1 XP_016869924.1 14 - 15 2150 93
XM_017014436.1 XP_016869925.1 14 - 15 2150 93
XM_017014437.1 XP_016869926.1 14 - 15 2150 93
XM_017014438.1 XP_016869927.1 14 - 15 2149 93
XM_017014439.1 XP_016869928.1 15 - 16 2312 93
XM_017014440.1 XP_016869929.1 12 - 13 1571 93
XM_017014441.1 XP_016869930.1 14 - 15 2149 93
XM_017014442.1 XP_016869931.1 14 - 15 2149 93
XM_017014443.1 XP_016869932.1 13 - 14 1632 93
XM_017014444.1 XP_016869933.1 3 - 4 351 93
GenBank mRNA AK128581.1 - 2 - 3 267 93
AK308284.1 - 14 - 15 1911 93
AY422473.1 - 14 - 15 2004 93
BC064849.1 - 1 - 2 210 93
BC144214.1 - 13 - 14 1805 93
BC150600.1 - 13 - 14 1805 93
CR936775.1 - 15 - 16 2282 93

View Full Product Details