The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002939.3 | 3 - 3 | 453 | 71 | |
GenBank mRNA | AK023827.1 | - | 4 - 4 | 667 | 71 |
BX648280.1 | - | 5 - 5 | 1125 | 71 |