The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001154458.2 | NP_001147930.1 | 5 - 6 | 756 | 70 |
NM_144605.4 | NP_653206.2 | 6 - 7 | 894 | 70 | |
XM_006720846.2 | XP_006720909.1 | 6 - 7 | 1030 | 70 | |
XM_011522379.2 | XP_011520681.1 | 5 - 6 | 543 | 70 | |
XM_017022938.1 | XP_016878427.1 | 5 - 6 | 1008 | 70 | |
GenBank mRNA | AK058139.1 | - | 6 - 7 | 720 | 70 |
AK098718.1 | - | 6 - 7 | 721 | 70 | |
BC024017.1 | - | 6 - 7 | 694 | 70 | |
BC035619.1 | - | 6 - 7 | 700 | 70 | |
DQ456996.1 | - | 5 - 6 | 582 | 70 | |
DQ517531.1 | - | 6 - 7 | 771 | 70 | |
EF620906.1 | - | 6 - 7 | 894 | 70 |