The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001891.3 | NP_001882.1 | 3 - 4 | 111 | 71 |
XM_017007760.1 | XP_016863249.1 | 4 - 5 | 189 | 71 | |
GenBank mRNA | BC069554.1 | - | 3 - 4 | 109 | 71 |
BC096194.3 | - | 3 - 4 | 109 | 71 | |
BC096195.1 | - | 3 - 4 | 109 | 71 | |
BC096196.3 | - | 3 - 4 | 112 | 71 | |
BC096197.3 | - | 3 - 4 | 109 | 71 | |
KF751868.1 | - | 3 - 4 | 95 | 71 |