Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SESN3
Entrez Gene ID
143686
Gene Name
sestrin 3
Gene Aliases
SEST3
Chromosome Location
Chr.11: 95165513 - 95232541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271594.1 NP_001258523.1 5 - 6 1060 119
NM_144665.3 NP_653266.2 6 - 7 1158 119
XM_011542614.1 XP_011540916.1 7 - 8 1262 119
GenBank mRNA AK096300.1 - 6 - 7 1158 119
AK302366.1 - 5 - 6 1060 119

View Full Product Details