The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018112.2 | NP_060582.1 | 3 - 4 | 591 | 125 |
XM_005252075.2 | XP_005252132.1 | 3 - 4 | 1038 | 125 | |
XM_005252076.2 | XP_005252133.1 | 3 - 4 | 572 | 125 | |
XM_005252077.3 | XP_005252134.1 | 3 - 4 | 572 | 125 | |
XM_011518829.1 | XP_011517131.1 | 3 - 4 | 572 | 125 | |
XM_011518831.2 | XP_011517133.1 | 3 - 4 | 572 | 125 | |
XM_011518833.2 | XP_011517135.1 | 4 - 5 | 723 | 125 | |
GenBank mRNA | AK001355.1 | - | 3 - 4 | 572 | 125 |
BC000049.2 | - | 3 - 4 | 547 | 125 | |
CR457258.1 | - | 3 - 4 | 455 | 125 |