The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161575.1 | NP_001155047.1 | 5 - 6 | 938 | 122 |
NM_018296.5 | NP_060766.5 | 8 - 9 | 1208 | 122 | |
XM_005256025.2 | XP_005256082.1 | 8 - 9 | 1208 | 122 | |
XM_005256027.2 | XP_005256084.1 | 8 - 9 | 1208 | 122 | |
XM_005256028.1 | XP_005256085.1 | 7 - 8 | 1080 | 122 | |
XM_011523200.1 | XP_011521502.1 | 8 - 9 | 1208 | 122 | |
XM_011523201.1 | XP_011521503.1 | 5 - 6 | 828 | 122 | |
XM_011523202.2 | XP_011521504.1 | 4 - 5 | 683 | 122 | |
XM_011523204.1 | XP_011521506.1 | 6 - 7 | 1007 | 122 | |
XM_011523205.1 | XP_011521507.1 | 7 - 8 | 1098 | 122 | |
XM_011523206.2 | XP_011521508.1 | 8 - 9 | 1208 | 122 | |
XM_017023400.1 | XP_016878889.1 | 8 - 9 | 1208 | 122 | |
GenBank mRNA | AK001866.1 | - | 5 - 6 | 987 | 122 |
AK292311.1 | - | 5 - 6 | 917 | 122 | |
AK301766.1 | - | 5 - 6 | 892 | 122 | |
AY313780.1 | - | 5 - 6 | 901 | 122 | |
AY963267.1 | - | 8 - 9 | 1208 | 122 | |
BC026156.2 | - | 8 - 9 | 1208 | 122 |