Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT9B
Entrez Gene ID
7484
Gene Name
Wnt family member 9B
Gene Aliases
WNT14B, WNT15
Chromosome Location
Chr.17: 46833201 - 46886730 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320458.1 NP_001307387.1 1 - 2 130 122
NM_003396.2 NP_003387.1 1 - 2 130 122
GenBank mRNA AB063483.1 - 1 - 2 114 122
AK127615.1 - 1 - 2 130 122
AY358217.1 - 1 - 2 150 122
BC064534.1 - 1 - 2 104 122

View Full Product Details