Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCBP1
Entrez Gene ID
4686
Gene Name
nuclear cap binding protein subunit 1
Gene Aliases
CBP80, NCBP, Sto1
Chromosome Location
Chr.9: 97633423 - 97673748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002486.4 NP_002477.1 9 - 10 1460 62
XM_011518715.2 XP_011517017.1 9 - 10 1459 62
XM_017014735.1 XP_016870224.1 9 - 10 1298 62
XM_017014736.1 XP_016870225.1 9 - 10 1011 62
XM_017014737.1 XP_016870226.1 9 - 10 999 62
GenBank mRNA AB209233.1 - 9 - 10 1040 62
AK312807.1 - 9 - 10 1460 62
BC001450.2 - 9 - 10 1064 62
D32002.1 - 9 - 10 1265 62
X80030.1 - 9 - 10 1031 62

View Full Product Details