Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location
Chr.20: 35771999 - 35950377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016436.4 NP_057520.2 7 - 8 1034 86
XM_005260421.2 XP_005260478.1 4 - 5 644 86
XM_011528843.1 XP_011527145.1 6 - 7 635 86
XM_011528844.1 XP_011527146.1 4 - 5 706 86
XM_017027864.1 XP_016883353.1 8 - 9 1116 86
XM_017027865.1 XP_016883354.1 8 - 9 1317 86
XM_017027866.1 XP_016883355.1 7 - 8 1237 86
XM_017027867.1 XP_016883356.1 7 - 8 1072 86
XM_017027868.1 XP_016883357.1 5 - 6 858 86
XM_017027869.1 XP_016883358.1 7 - 8 1031 86
XM_017027870.1 XP_016883359.1 7 - 8 1037 86
XM_017027871.1 XP_016883360.1 8 - 9 1117 86
GenBank mRNA AF220416.1 - 8 - 9 937 86
AF258787.1 - 7 - 8 1020 86
AF348207.1 - 6 - 7 959 86
AK054597.1 - 8 - 9 1291 86
AK314503.1 - 7 - 8 998 86
AY027523.1 - 6 - 7 959 86
AY134747.1 - 7 - 8 1011 86
BC006415.1 - 6 - 7 894 86
BC016948.1 - 7 - 8 987 86
BC048210.1 - 8 - 9 1067 86
BC080598.1 - 7 - 8 998 86
BC093405.1 - 7 - 8 1034 86
BC150178.1 - 7 - 8 1000 86
L09749.1 - 6 - 7 914 86

View Full Product Details