Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF71
Entrez Gene ID
58491
Gene Name
zinc finger protein 71
Gene Aliases
EZFIT
Chromosome Location
Chr.19: 56595259 - 56624481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527193.2 XP_011525495.1 2 - 3 240 101
XM_017027078.1 XP_016882567.1 2 - 3 150 101
XM_017027079.1 XP_016882568.1 2 - 3 2020 101

View Full Product Details