The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014938.5 | NP_055753.3 | 2 - 3 | 639 | 68 |
XM_006719290.3 | XP_006719353.1 | 2 - 3 | 623 | 68 | |
XM_006719291.3 | XP_006719354.1 | 2 - 3 | 623 | 68 | |
XM_006719292.3 | XP_006719355.1 | 2 - 3 | 623 | 68 | |
XM_006719293.3 | XP_006719356.1 | 2 - 3 | 623 | 68 | |
GenBank mRNA | AF312918.1 | - | 2 - 3 | 513 | 68 |
BC151841.1 | - | 2 - 3 | 621 | 68 | |
BM461435.1 | - | 2 - 3 | 675 | 68 |